siRNA Details
Gene Symbol: RPS15A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001019.4 | NP_001010.2 | 4 | 351 | ||
NM_001030009.1 | NP_001025180.1 | 4 | 285 | |||
GenBank mRNA | AB062400.1 | - | 1 | 267 | ||
AK023088.1 | - | 5 | 380 | |||
AK130967.1 | - | 4 | 285 | |||
AK291988.1 | - | 4 | 351 | |||
AK311976.1 | - | 1 | 285 | |||
AY208299.1 | - | 3 | 261 | |||
BC001697.2 | - | 4 | 320 | 2967511 | ||
BC030569.1 | - | 3 | 283 | 5580542 | ||
BC046113.1 | - | 1 | 268 | 5736078 | ||
BC105273.1 | - | 1 | 291 | 3839617 | ||
BC105292.1 | - | 4 | 320 | 6649512 | ||
BF678907.1 | - | 4 | 352 | |||
BG035607.1 | - | 338 | ||||
X84407.1 | - | 1 | 255 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
