siRNA Details
Gene Symbol: RRP7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015703.4 | NP_056518.2 | 2 | 216 | ||
GenBank mRNA | AK314650.1 | - | 2 | 182 | ||
AY039240.1 | - | 1 | 88 | |||
BC031838.1 | - | 1 | 131 | |||
BC035992.1 | - | 2 | 195 | |||
BC041639.1 | - | 2 | 174 | |||
BC042335.1 | - | 2 | 177 | |||
BC073834.1 | - | 2 | 189 | 5533064 | ||
BC121118.1 | - | 2 | 199 | 40120513 | ||
BC121119.1 | - | 2 | 199 | 40120514 | ||
BP314581.1 | - | 2 | 216 |
Gene Symbol: RRP7BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002184.2 | 2 | 189 | |||
GenBank mRNA | AK098201.1 | - | 2 | 216 | ||
AL157851.1 | - | 2 | 174 | |||
BC007736.2 | - | 2 | 189 | |||
BC014647.1 | - | 2 | 187 | |||
BC143340.1 | - | 2 | 277 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
