siRNA Details
Gene Symbol: RRP7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015703.4 | NP_056518.2 | 4, 5 | 500 | ||
GenBank mRNA | AK092492.1 | - | 2, 3 | 1026 | ||
AK314650.1 | - | 4, 5 | 466 | |||
AY039240.1 | - | 3, 4 | 372 | |||
BC031838.1 | - | 3, 4 | 415 | |||
BC035992.1 | - | 4, 5 | 479 | |||
BC041639.1 | - | 4, 5 | 458 | |||
BC042335.1 | - | 4, 5 | 461 | |||
BC073834.1 | - | 4, 5 | 473 | 5533064 | ||
BC121118.1 | - | 4, 5 | 483 | 40120513 | ||
BC121119.1 | - | 4, 5 | 483 | 40120514 | ||
BP314581.1 | - | 4, 5 | 500 |
Gene Symbol: RRP7BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002184.2 | 4, 5 | 466 | |||
GenBank mRNA | BC007736.2 | - | 4, 5 | 466 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
