siRNA Details
Gene Symbol: RRP7BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002184.2 | 7 | 794 | |||
GenBank mRNA | AK074492.1 | - | 5 | 555 | ||
AK098201.1 | - | 7 | 821 | |||
AL157851.1 | - | 7 | 779 | |||
BC007736.2 | - | 7 | 794 | |||
BC014647.1 | - | 7 | 792 | |||
BC143340.1 | - | 7 | 882 |
Gene Symbol: RRP7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015703.4 | NP_056518.2 | 7 | 831 | ||
GenBank mRNA | AK092492.1 | - | 5 | 1357 | ||
AK314650.1 | - | 7 | 797 | |||
AY039240.1 | - | 6 | 703 | |||
BC031838.1 | - | 6 | 746 | |||
BC035992.1 | - | 7 | 810 | |||
BC041639.1 | - | 7 | 789 | |||
BC042335.1 | - | 7 | 792 | |||
BC063537.1 | - | 3 | 327 | |||
BC073834.1 | - | 7 | 804 | 5533064 | ||
BC121118.1 | - | 7 | 814 | 40120513 | ||
BC121119.1 | - | 7 | 814 | 40120514 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
