siRNA Details
Gene Symbol: SUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006713.3 | NP_006704.3 | 2 | 190 | ||
XM_011513944.2 | XP_011512246.1 | 3 | 443 | |||
XM_017008986.1 | XP_016864475.1 | 3 | 286 | |||
XM_017008987.1 | XP_016864476.1 | 3 | 163 | |||
GenBank mRNA | AB209285.1 | - | 2 | 85 | ||
AK294124.1 | - | 2 | 132 | |||
AK307268.1 | - | 2 | 122 | |||
AK311754.1 | - | 2 | 132 | |||
BC009610.1 | - | 2 | 118 | 4297490 | ||
BC010537.1 | - | 2 | 93 | 3457167 | ||
BC018189.1 | - | 2 | 112 | 3881373 | ||
BC022339.1 | - | 2 | 123 | 4134117 | ||
BC063402.1 | - | 2 | 106 | |||
BC071655.1 | - | 2 | 107 | |||
BC105972.1 | - | 2 | 88 | 3907729 | ||
BX472106.1 | - | 2 | 191 | |||
CR456902.1 | - | 1 | 62 | |||
U12979.1 | - | 1 | 62 | |||
X79805.1 | - | 1 | 62 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
