siRNA Details
Gene Symbol: DTX2P1-UPK3BP1-PMS2P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023383.1 | 8, 9 | 1330 | |||
GenBank mRNA | U38980.1 | - | 8, 9 | 1216 |
Gene Symbol: PMS2P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC047753.1 | - | 3, 4 | 265 |
Gene Symbol: PMS2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003614.3 | 3, 4 | 289 | |||
GenBank mRNA | DA215155.1 | - | 3, 4 | 276 |
Gene Symbol: PMS2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027776.2 | 3, 4 | 314 | |||
GenBank mRNA | AK309660.1 | - | 3, 4 | 262 | ||
BF984173.1 | - | 3, 4 | 313 | |||
D38439.1 | - | 2, 3 | 226 | |||
D38501.1 | - | 2, 3 | 226 |
Gene Symbol: PMS2P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028058.1 | 2, 3 | 705 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
