siRNA Details
Gene Symbol: PMS2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027775.2 | 5 | 574 | |||
NR_027776.2 | 4 | 471 | ||||
NR_027777.2 | 3 | 384 | ||||
GenBank mRNA | AK309660.1 | - | 4 | 419 | ||
BC052588.1 | - | 3 | 387 | |||
BF984173.1 | - | 4 | 470 | |||
D38439.1 | - | 3 | 378 | |||
D38501.1 | - | 3 | 378 |
Gene Symbol: DTX2P1-UPK3BP1-PMS2P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023383.1 | 9 | 1487 | |||
GenBank mRNA | U38980.1 | - | 9 | 1373 |
Gene Symbol: LOC441259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC075816.1 | - | 5 | 529 |
Gene Symbol: PMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000535.6 | NP_000526.2 | 5 | 605 | ||
NM_001322003.1 | NP_001308932.1 | 5 | 600 | |||
NM_001322004.1 | NP_001308933.1 | 4 | 465 | |||
NM_001322005.1 | NP_001308934.1 | 5 | 717 | |||
NM_001322006.1 | NP_001308935.1 | 5 | 605 | |||
NM_001322007.1 | NP_001308936.1 | 4 | 502 | |||
NM_001322008.1 | NP_001308937.1 | 3 | 362 | |||
NM_001322009.1 | NP_001308938.1 | 5 | 712 | |||
NM_001322010.1 | NP_001308939.1 | 4 | 465 | |||
NM_001322011.1 | NP_001308940.1 | 5 | 605 | |||
NM_001322012.1 | NP_001308941.1 | 5 | 600 | |||
NM_001322013.1 | NP_001308942.1 | 5 | 600 | |||
NM_001322014.1 | NP_001308943.1 | 5 | 605 | |||
NM_001322015.1 | NP_001308944.1 | 5 | 775 | |||
NR_136154.1 | 5 | 605 | ||||
XM_017012342.1 | XP_016867831.1 | 5 | 579 | |||
GenBank mRNA | AB103082.1 | - | 5 | 518 | ||
AB103083.1 | - | 5 | 518 | |||
AB103085.1 | - | 4 | 518 | |||
AB103086.1 | - | 2 | 200 | |||
AK294661.1 | - | 4 | 455 | |||
AK312390.1 | - | 5 | 554 | |||
BC008400.1 | - | 5 | 563 | |||
BC031832.1 | - | 5 | 569 | |||
BC093921.1 | - | 5 | 540 | 7939766 | ||
BP251389.1 | - | 1 | 75 | |||
CR981250.1 | - | 5 | 506 | |||
U14658.1 | - | 5 | 520 |
Gene Symbol: PMS2P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC047753.1 | - | 4 | 424 |
Gene Symbol: PMS2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003614.3 | 4 | 448 |
Gene Symbol: PMS2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028059.1 | 8 | 1361 | |||
GenBank mRNA | D38437.1 | - | 4 | 509 | ||
D38498.1 | - | 4 | 509 |
Gene Symbol: PMS2P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_022007.2 | 4 | 459 | |||
NR_046297.1 | 4 | 459 | ||||
NR_046299.1 | 4 | 459 | ||||
GenBank mRNA | BC029368.1 | - | 4 | 436 | ||
BC029419.1 | - | 4 | 424 | |||
CB141754.1 | - | 4 | 424 | |||
D38438.1 | - | 4 | 428 | |||
D38500.1 | - | 4 | 428 | |||
DA885243.1 | - | 4 | 460 |
Gene Symbol: PMS2P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB017005.1 | - | 1 | 124 |
Gene Symbol: PMS2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130940.1 | 5 | 602 | |||
GenBank mRNA | AB017006.1 | - | 5 | 695 |
Gene Symbol: PMS2P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028058.1 | 3 | 862 | |||
GenBank mRNA | BM845848.1 | - | 4 | 403 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
