siRNA Details
Gene Symbol: FKBP9P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003949.1 | 5 | 637 | |||
NR_027339.1 | 3 | 741 | ||||
NR_027340.1 | 2 | 370 | ||||
NR_027342.1 | 3 | 651 | ||||
GenBank mRNA | AK289438.1 | - | 2 | 326 | ||
AK312981.1 | - | 2 | 370 | |||
BC011872.2 | - | 3 | 651 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
