siRNA Details
Gene Symbol: FKBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284341.1 | NP_001271270.1 | 11 | 2451 | ||
NM_001284343.1 | NP_001271272.1 | 7 | 1529 | |||
NM_007270.4 | NP_009201.2 | 10 | 2292 | |||
GenBank mRNA | AB209352.1 | - | 3 | 4011 | ||
AK075331.1 | - | 10 | 2256 | |||
AK128597.1 | - | 11 | 2233 | |||
AK293954.1 | - | 9 | 1866 | |||
AK304643.1 | - | 7 | 1367 | |||
AK316290.1 | - | 5 | 1182 | |||
AL050187.1 | - | 2 | 622 | |||
BC007443.2 | - | 4 | 1008 | |||
BC064418.1 | - | 5 | 1215 | |||
BC072422.1 | - | 4 | 959 | |||
BC099918.1 | - | 3 | 854 |
Gene Symbol: FKBP9P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003949.1 | 6 | 1271 | |||
NR_027339.1 | 4 | 1375 | ||||
NR_027340.1 | 3 | 1004 | ||||
NR_027342.1 | 4 | 1285 | ||||
GenBank mRNA | BC011872.2 | - | 4 | 1285 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
