siRNA Details
Gene Symbol: RPL39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000.3 | NP_000991.1 | 2 | 135 | ||
GenBank mRNA | AK098280.1 | - | 1 | 1633 | ||
BC001019.2 | - | 1 | 87 | 3343512 | ||
BC070205.1 | - | 1 | 105 | 6732169 | ||
BX476041.1 | - | 2 | 136 | |||
D79205.1 | - | 2 | 89 | |||
U57846.1 | - | 2 | 70 |
Gene Symbol: LMNTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DW009862.1 | - | 5 | 549 |
Gene Symbol: RPL39L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052969.1 | NP_443201.1 | 3 | 407 | ||
GenBank mRNA | AB063610.1 | - | 3 | 399 | ||
AF548529.1 | - | 1 | 52 | |||
BC012328.1 | - | 3 | 407 | 4555759 | ||
L05096.1 | - | 3 | 353 |
Gene Symbol: RPL39P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209077.1 | - | 2 | 264 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
