siRNA Details
Gene Symbol: FAM153B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265615.1 | NP_001252544.1 | 6 | 261 | ||
GenBank mRNA | BC028606.1 | - | 7 | 502 | 5266126 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
