siRNA Details
Gene Symbol: FAM153B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265615.1 | NP_001252544.1 | 12 | 576 | ||
GenBank mRNA | BC028606.1 | - | 13 | 817 | 5266126 | |
CD580216.2 | - | 5 | 219 |
Gene Symbol: LOC102724062 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546026.2 | XP_011544328.1 | 6 | 426 |
Gene Symbol: LOC107987382 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030119.1 | XP_016885608.1 | 5 | 323 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
