siRNA Details
Gene Symbol: IDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000202.7 | NP_000193.1 | 3 | 534 | ||
NM_001166550.3 | NP_001160022.1 | 3 | 490 | |||
NM_006123.4 | NP_006114.1 | 3 | 534 | |||
NR_104128.1 | 3 | 534 | ||||
GenBank mRNA | AF050145.1 | - | 3 | 335 | ||
AK294541.1 | - | 3 | 445 | |||
BC006170.1 | - | 3 | 392 | 4025980 | ||
BX647357.1 | - | 3 | 388 | |||
DA325538.1 | - | 3 | 534 | |||
L40586.1 | - | 3 | 444 | |||
M58342.1 | - | 3 | 441 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
