siRNA Details
Gene Symbol: MLLT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005937.3 | NP_005928.2 | 16 | 2442 | ||
XM_006721911.2 | XP_006721974.1 | 16 | 2364 | |||
XM_017024648.1 | XP_016880137.1 | 11 | 1967 | |||
GenBank mRNA | AB209633.1 | - | 16 | 2417 | ||
AK027133.1 | - | 5 | 560 | |||
AL133659.1 | - | 4 | 1421 | |||
BC007237.1 | - | 8 | 1393 | |||
BC007859.2 | - | 6 | 793 | |||
U07932.1 | - | 16 | 2429 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
