siRNA Details
Gene Symbol: ANKRD18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147195.2 | NP_671728.2 | 15 | 2945 | ||
XM_006716749.3 | XP_006716812.1 | 16 | 3222 | |||
XM_011517836.2 | XP_011516138.1 | 16 | 3223 | |||
XM_011517837.2 | XP_011516139.1 | 16 | 3223 | |||
XM_011517839.2 | XP_011516141.1 | 15 | 3167 | |||
XM_017014576.1 | XP_016870065.1 | 17 | 3354 | |||
GenBank mRNA | AB095935.1 | - | 15 | 2890 | ||
AK093059.1 | - | 2 | 281 | |||
AY550933.1 | - | 2 | 230 | |||
BC131497.1 | - | 2 | 277 | |||
BC152434.1 | - | 15 | 2913 | 8862553 | ||
DB066861.1 | - | 2 | 334 |
Gene Symbol: ANKRD18B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244752.1 | NP_001231681.1 | 15 | 2968 | ||
XM_011517881.2 | XP_011516183.1 | 16 | 3035 | |||
XM_011517883.2 | XP_011516185.1 | 15 | 2978 | |||
XM_011517885.2 | XP_011516187.1 | 10 | 2123 | |||
XM_017014728.1 | XP_016870217.1 | 10 | 2120 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
