siRNA Details
Gene Symbol: ANKRD18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147195.2 | NP_671728.2 | 14 | 2800 | ||
XM_006716749.3 | XP_006716812.1 | 15 | 3077 | |||
XM_011517836.2 | XP_011516138.1 | 15 | 3078 | |||
XM_011517837.2 | XP_011516139.1 | 15 | 3078 | |||
XM_011517839.2 | XP_011516141.1 | 14 | 3022 | |||
XM_017014576.1 | XP_016870065.1 | 16 | 3209 | |||
GenBank mRNA | AB095935.1 | - | 14 | 2745 | ||
AK093059.1 | - | 1 | 136 | |||
AY550933.1 | - | 1 | 85 | |||
BC131497.1 | - | 1 | 132 | |||
BC152434.1 | - | 14 | 2768 | 8862553 | ||
DB066861.1 | - | 1 | 189 |
Gene Symbol: ANKRD20A8P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003366.2 | 18 | 3107 |
Gene Symbol: LOC644249 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015390.1 | XP_016870879.1 | 4 | 681 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
