siRNA Details
Gene Symbol: CCDC177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271507.1 | NP_001258436.1 | 2 | 3052 | ||
GenBank mRNA | AF284217.1 | - | 1 | 261 | ||
AK312175.1 | - | 1 | 375 | |||
BC105025.1 | - | 1 | 184 | |||
BC105027.1 | - | 1 | 184 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
