siRNA Details
Gene Symbol: RGPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078170.2 | NP_001071638.2 | 9 | 1140 | ||
XM_017004845.1 | XP_016860334.1 | 13 | 1591 | |||
XM_017004846.1 | XP_016860335.1 | 9 | 1262 | |||
XM_017004847.1 | XP_016860336.1 | 9 | 1259 | |||
XM_017004848.1 | XP_016860337.1 | 13 | 1591 | |||
XM_017004849.1 | XP_016860338.1 | 13 | 1591 | |||
XM_017004850.1 | XP_016860339.1 | 13 | 1591 | |||
GenBank mRNA | AK302429.1 | - | 9 | 1140 |
Gene Symbol: RANBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006267.4 | NP_006258.3 | 9 | 1228 | ||
XM_005264002.2 | XP_005264059.1 | 9 | 1263 | |||
XM_005264003.2 | XP_005264060.1 | 9 | 1191 | |||
XM_005264004.2 | XP_005264061.1 | 9 | 1191 | |||
XM_005264005.4 | XP_005264062.1 | 9 | 1251 | |||
XM_005264007.2 | XP_005264064.1 | 9 | 1191 | |||
XM_011511575.2 | XP_011509877.1 | 9 | 1166 | |||
XM_011511576.2 | XP_011509878.1 | 9 | 1191 | |||
XM_011511578.2 | XP_011509880.1 | 9 | 1233 | |||
XM_017004623.1 | XP_016860112.1 | 9 | 1191 | |||
XM_017004624.1 | XP_016860113.1 | 9 | 1191 | |||
XM_017004625.1 | XP_016860114.1 | 9 | 1263 | |||
GenBank mRNA | AB209483.1 | - | 6 | 844 | ||
D42063.1 | - | 9 | 1229 |
Gene Symbol: RGPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024457.3 | NP_001019628.3 | 9 | 1094 | ||
XM_011532845.2 | XP_011531147.1 | 10 | 1293 | |||
XM_011532849.2 | XP_011531151.1 | 10 | 1293 | |||
XM_011532850.1 | XP_011531152.1 | 10 | 1293 | |||
XM_011532851.2 | XP_011531153.1 | 10 | 1293 | |||
XM_011532852.2 | XP_011531154.1 | 10 | 1293 | |||
XM_017004103.1 | XP_016859592.1 | 10 | 1293 | |||
XM_017004104.1 | XP_016859593.1 | 9 | 1410 | |||
XM_017004105.1 | XP_016859594.1 | 9 | 1258 | |||
XM_017004106.1 | XP_016859595.1 | 9 | 1377 | |||
XM_017004107.1 | XP_016859596.1 | 10 | 1293 | |||
GenBank mRNA | AF237700.2 | - | 1 | 43 | ||
CR749330.1 | - | 9 | 1094 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
