siRNA Details
Gene Symbol: WASH2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024077.2 | 9 | 1363 | |||
GenBank mRNA | AK292759.1 | - | 8 | 970 | ||
AK308574.1 | - | 8 | 1132 | |||
AL137714.1 | - | 7 | 2145 | |||
AY217347.1 | - | 8 | 1790 | |||
AY341936.1 | - | 4 | 496 | |||
AY341937.1 | - | 4 | 496 | |||
BC015400.1 | - | 1 | 55 | |||
BC073913.1 | - | 4 | 759 | |||
BC082240.1 | - | 6 | 2058 | |||
BC093083.1 | - | 4 | 772 | |||
BC106020.1 | - | 6 | 800 | |||
BX537637.1 | - | 9 | 1364 |
Gene Symbol: LOC100288778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130745.1 | 9 | 1242 | |||
GenBank mRNA | AW881719.1 | - | 1 | 419 | ||
BC063682.1 | - | 5 | 588 |
Gene Symbol: WASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182905.4 | NP_878908.4 | 9 | 1242 | ||
XM_011517658.2 | XP_011515960.1 | 9 | 1359 | |||
XM_011517659.2 | XP_011515961.1 | 9 | 1141 | |||
XM_011517660.2 | XP_011515962.1 | 9 | 1138 | |||
XM_011517662.2 | XP_011515964.1 | 9 | 2645 | |||
XM_011517663.2 | XP_011515965.1 | 8 | 2494 | |||
XM_011517664.2 | XP_011515966.1 | 8 | 2491 | |||
XM_011517665.2 | XP_011515967.1 | 8 | 1040 | |||
XM_011517666.2 | XP_011515968.1 | 8 | 1039 | |||
XM_017014169.1 | XP_016869658.1 | 9 | 1258 | |||
XM_017014170.1 | XP_016869659.1 | 9 | 1321 | |||
XM_017014171.1 | XP_016869660.1 | 7 | 933 | |||
XM_017014172.1 | XP_016869661.1 | 7 | 2511 | |||
GenBank mRNA | AK024481.1 | - | 2 | 2751 | ||
AK289708.1 | - | 9 | 1254 | |||
BC094860.1 | - | 1 | 283 | |||
CN404407.1 | - | 2 | 139 |
Gene Symbol: WASH3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003659.2 | 9 | 1916 | |||
GenBank mRNA | AK056232.1 | - | 9 | 1761 | ||
AK057951.1 | - | 11 | 1366 | |||
AK095183.1 | - | 8 | 1362 | |||
AK290087.1 | - | 7 | 982 | |||
AK291582.1 | - | 8 | 1377 | |||
BC048328.1 | - | 8 | 1125 | |||
BC063555.1 | - | 8 | 1551 | |||
BC110991.1 | - | 4 | 525 | |||
BQ935819.1 | - | 4 | 528 |
Gene Symbol: WASH6P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC029274.1 | - | 2 | 151 |
Gene Symbol: WASH7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024540.1 | 9 | 1136 | |||
GenBank mRNA | AK092583.1 | - | 7 | 1671 | ||
AK302089.1 | - | 9 | 1191 | |||
AK308566.1 | - | 7 | 874 | |||
BC063470.1 | - | 6 | 1050 | |||
BC094698.1 | - | 6 | 1912 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
