siRNA Details
Gene Symbol: WASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182905.4 | NP_878908.4 | 8, 9 | 1184 | ||
XM_011517658.2 | XP_011515960.1 | 8, 9 | 1301 | |||
XM_011517659.2 | XP_011515961.1 | 8, 9 | 1083 | |||
XM_011517660.2 | XP_011515962.1 | 8, 9 | 1080 | |||
XM_011517662.2 | XP_011515964.1 | 8, 9 | 2587 | |||
XM_011517663.2 | XP_011515965.1 | 7, 8 | 2436 | |||
XM_011517664.2 | XP_011515966.1 | 7, 8 | 2433 | |||
XM_011517665.2 | XP_011515967.1 | 7, 8 | 982 | |||
XM_011517666.2 | XP_011515968.1 | 7, 8 | 981 | |||
XM_017014169.1 | XP_016869658.1 | 8, 9 | 1200 | |||
XM_017014170.1 | XP_016869659.1 | 8, 9 | 1263 | |||
XM_017014171.1 | XP_016869660.1 | 6, 7 | 875 | |||
XM_017014172.1 | XP_016869661.1 | 6, 7 | 2453 | |||
GenBank mRNA | CN404407.1 | - | 1, 2 | 81 |
Gene Symbol: WASH3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK057951.1 | - | 10, 11 | 1308 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
