siRNA Details
Gene Symbol: WASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182905.4 | NP_878908.4 | 8 | 1158 | ||
XM_011517658.2 | XP_011515960.1 | 8 | 1275 | |||
XM_011517659.2 | XP_011515961.1 | 8 | 1057 | |||
XM_011517660.2 | XP_011515962.1 | 8 | 1054 | |||
XM_011517662.2 | XP_011515964.1 | 8 | 2561 | |||
XM_011517663.2 | XP_011515965.1 | 7 | 2410 | |||
XM_011517664.2 | XP_011515966.1 | 7 | 2407 | |||
XM_011517665.2 | XP_011515967.1 | 7 | 956 | |||
XM_011517666.2 | XP_011515968.1 | 7 | 955 | |||
XM_011517671.2 | XP_011515973.2 | 7 | 1135 | |||
XM_017014169.1 | XP_016869658.1 | 8 | 1174 | |||
XM_017014170.1 | XP_016869659.1 | 8 | 1237 | |||
XM_017014171.1 | XP_016869660.1 | 6 | 849 | |||
XM_017014172.1 | XP_016869661.1 | 6 | 2427 | |||
XM_017014173.1 | XP_016869662.1 | 7 | 1139 | |||
GenBank mRNA | AK024481.1 | - | 2 | 2008 | ||
CN404407.1 | - | 1 | 55 |
Gene Symbol: LOC100288778 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130745.1 | 8 | 1158 | |||
GenBank mRNA | BC063682.1 | - | 4 | 504 |
Gene Symbol: WASH2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024077.2 | 8 | 1279 | |||
GenBank mRNA | AK292759.1 | - | 7 | 886 | ||
AK308574.1 | - | 7 | 968 | |||
AL137714.1 | - | 7 | 1405 | |||
AY217347.1 | - | 7 | 1706 | |||
AY341936.1 | - | 3 | 412 | |||
AY341937.1 | - | 3 | 412 | |||
BC073913.1 | - | 3 | 675 | |||
BC082240.1 | - | 5 | 1974 | |||
BC093083.1 | - | 3 | 688 | |||
BC106020.1 | - | 5 | 716 | |||
BX537637.1 | - | 8 | 1280 |
Gene Symbol: WASH3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003659.2 | 8 | 1832 | |||
GenBank mRNA | AK056232.1 | - | 8 | 1677 | ||
AK057951.1 | - | 10 | 1282 | |||
AK095183.1 | - | 7 | 1278 | |||
AK290087.1 | - | 6 | 898 | |||
AK291582.1 | - | 7 | 1293 | |||
BC048328.1 | - | 7 | 1041 | |||
BC063555.1 | - | 7 | 1467 | |||
BC110991.1 | - | 3 | 441 | |||
BQ935819.1 | - | 3 | 444 |
Gene Symbol: WASH6P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC029274.1 | - | 1 | 67 |
Gene Symbol: WASH7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024540.1 | 8 | 1053 | |||
GenBank mRNA | AK092583.1 | - | 7 | 928 | ||
AK296182.1 | - | 9 | 1189 | |||
AK302089.1 | - | 8 | 1107 | |||
AK308566.1 | - | 6 | 790 | |||
BC063470.1 | - | 5 | 966 | |||
BC094698.1 | - | 5 | 1828 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
