siRNA Details
Gene Symbol: DUSP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004420.2 | NP_004411.2 | 2 | 250 | ||
XM_011519932.2 | XP_011518234.1 | 2 | 1277 | |||
XM_011519933.2 | XP_011518235.1 | 2 | 307 | |||
GenBank mRNA | BC038231.1 | - | 1 | 253 | 5547764 | |
BC045110.1 | - | 1 | 123 | 5478677 | ||
U27193.1 | - | 2 | 256 |
Gene Symbol: GLUD1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY661565.1 | - | 1 | 120 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
