siRNA Details
Gene Symbol: ST13P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF512499.1 | - | 1 | 357 |
Gene Symbol: LOC107986532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017011596.1 | XP_016867085.1 | 1 | 305 |
Gene Symbol: ST13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278589.1 | NP_001265518.1 | 4 | 722 | ||
NM_003932.4 | NP_003923.2 | 4 | 752 | |||
GenBank mRNA | AK130770.1 | - | 4 | 385 | ||
AK303530.1 | - | 4 | 353 | |||
AK312399.1 | - | 4 | 366 | |||
AK316155.1 | - | 4 | 271 | |||
AY513286.1 | - | 4 | 350 | |||
AY826824.1 | - | 4 | 360 | |||
AY826825.1 | - | 4 | 360 | |||
BC052982.1 | - | 4 | 374 | 6046993 | ||
BC071629.1 | - | 4 | 352 | 4536907 | ||
BC101206.1 | - | 4 | 308 | |||
BC101207.2 | - | 4 | 308 | |||
BC101208.1 | - | 4 | 308 | |||
BC107148.1 | - | 4 | 309 | 40031944 | ||
BC121107.1 | - | 4 | 360 | |||
BC121108.1 | - | 4 | 360 | |||
BC139724.1 | - | 4 | 309 | 40031942 | ||
BG706239.1 | - | 432 | ||||
CR456586.1 | - | 4 | 392 | |||
U17714.1 | - | 4 | 358 | |||
U28918.1 | - | 4 | 377 |
Gene Symbol: ST13P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002183.1 | 1 | 343 | |||
GenBank mRNA | AF539468.1 | - | 1 | 343 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
