siRNA Details
Gene Symbol: ST13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278589.1 | NP_001265518.1 | 11 | 1374 | ||
NM_003932.4 | NP_003923.2 | 11 | 1404 | |||
GenBank mRNA | AK130770.1 | - | 11 | 1037 | ||
AK303530.1 | - | 11 | 1005 | |||
AK312399.1 | - | 11 | 1019 | |||
AK316155.1 | - | 11 | 923 | |||
AY513286.1 | - | 11 | 1002 | |||
AY826824.1 | - | 11 | 1013 | |||
AY826825.1 | - | 11 | 1013 | |||
BC052982.1 | - | 11 | 1026 | 6046993 | ||
BC071629.1 | - | 11 | 1004 | 4536907 | ||
BC139724.1 | - | 11 | 961 | 40031942 | ||
CR456586.1 | - | 11 | 1044 | |||
U17714.1 | - | 11 | 1010 | |||
U28918.1 | - | 11 | 1029 |
Gene Symbol: ST13P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002183.1 | 1 | 995 | |||
GenBank mRNA | AF539468.1 | - | 1 | 995 |
Gene Symbol: ST13P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF512499.1 | - | 1 | 1009 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
