siRNA Details
Gene Symbol: ST13P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002183.1 | 1 | 1044 | |||
GenBank mRNA | AF539468.1 | - | 1 | 1044 |
Gene Symbol: ST13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278589.1 | NP_001265518.1 | 11, 12 | 1423 | ||
NM_003932.4 | NP_003923.2 | 11, 12 | 1453 | |||
GenBank mRNA | AK130770.1 | - | 11, 12 | 1086 | ||
AK303530.1 | - | 11, 12 | 1054 | |||
AK312399.1 | - | 11, 12 | 1068 | |||
AK316155.1 | - | 11, 12 | 972 | |||
AY513286.1 | - | 11, 12 | 1051 | |||
AY826824.1 | - | 11, 12 | 1062 | |||
AY826825.1 | - | 11, 12 | 1062 | |||
BC052982.1 | - | 11, 12 | 1075 | 6046993 | ||
BC071629.1 | - | 11, 12 | 1053 | 4536907 | ||
BC107148.1 | - | 9, 10 | 702 | 40031944 | ||
BC139724.1 | - | 11, 12 | 1010 | 40031942 | ||
CR456586.1 | - | 11, 12 | 1093 | |||
U17714.1 | - | 11, 12 | 1059 | |||
U28918.1 | - | 11, 12 | 1078 |
Gene Symbol: ST13P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF512499.1 | - | 1 | 1058 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
