siRNA Details
Gene Symbol: ANAPC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002244.2 | NP_001002244.1 | 4 | 883 | ||
NM_001002245.2 | NP_001002245.1 | 3 | 552 | |||
NM_001002246.2 | NP_001002246.1 | 4 | 615 | |||
NM_001002247.2 | NP_001002247.1 | 3 | 560 | |||
NM_001002248.2 | NP_001002248.1 | 4 | 623 | |||
NM_001002249.2 | NP_001002249.1 | 4 | 674 | |||
NM_001289414.1 | NP_001276343.1 | 4 | 556 | |||
NM_001289415.1 | NP_001276344.1 | 3 | 582 | |||
NM_001289416.1 | NP_001276345.1 | 3 | 611 | |||
NM_001289417.1 | NP_001276346.1 | 5 | 707 | |||
NM_001289418.1 | NP_001276347.1 | 3 | 590 | |||
NM_001289419.1 | NP_001276348.1 | 4 | 661 | |||
NM_001289420.1 | NP_001276349.1 | 5 | 760 | |||
NM_016476.11 | NP_057560.8 | 4 | 645 | |||
GenBank mRNA | AF151048.1 | - | 4 | 590 | ||
AF247565.1 | - | 3 | 490 | |||
AF247789.1 | - | 4 | 568 | |||
AK091360.1 | - | 1 | 2110 | |||
AK308091.1 | - | 2 | 466 | |||
AW965775.1 | - | 4 | 555 | |||
BC000607.1 | - | 4 | 859 | 3346039 | ||
BC066308.1 | - | 4 | 659 | |||
BC095454.1 | - | 4 | 574 | |||
BC104641.1 | - | 3 | 508 | |||
BC171892.1 | - | 4 | 557 | |||
BC171898.1 | - | 3 | 524 | |||
BC171899.1 | - | 4 | 565 | |||
BC171900.1 | - | 3 | 494 | |||
BG707824.1 | - | 4 | 606 | |||
BU554167.1 | - | 4 | 625 | |||
BU556841.1 | - | 4 | 589 | |||
BY796995.2 | - | 3 | 475 | |||
DB573049.1 | - | 2 | 426 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
