siRNA Details
Gene Symbol: ANAPC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002244.2 | NP_001002244.1 | 4 | 1028 | ||
NM_001002245.2 | NP_001002245.1 | 3 | 697 | |||
NM_001002246.2 | NP_001002246.1 | 4 | 760 | |||
NM_001002247.2 | NP_001002247.1 | 3 | 705 | |||
NM_001002248.2 | NP_001002248.1 | 4 | 768 | |||
NM_001002249.2 | NP_001002249.1 | 4 | 819 | |||
NM_001289414.1 | NP_001276343.1 | 4 | 701 | |||
NM_001289415.1 | NP_001276344.1 | 3 | 727 | |||
NM_001289416.1 | NP_001276345.1 | 3 | 756 | |||
NM_001289417.1 | NP_001276346.1 | 5 | 852 | |||
NM_001289418.1 | NP_001276347.1 | 3 | 735 | |||
NM_001289419.1 | NP_001276348.1 | 4 | 806 | |||
NM_001289420.1 | NP_001276349.1 | 5 | 905 | |||
NM_016476.11 | NP_057560.8 | 4 | 790 | |||
GenBank mRNA | AK308091.1 | - | 2 | 611 | ||
BU554167.1 | - | 4 | 770 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
