siRNA Details
Gene Symbol: BOLA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035505.1 | NP_001030582.1 | 2 | 156 | ||
NM_212552.2 | NP_997717.2 | 2 | 156 | |||
GenBank mRNA | BC028282.1 | - | 2 | 116 | ||
BC042036.1 | - | 2 | 152 | |||
BU600037.1 | - | 161 | ||||
DQ225187.1 | - | 2 | 118 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
