siRNA Details
Gene Symbol: BOLA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035505.1 | NP_001030582.1 | 3 | 219 | ||
NM_212552.2 | NP_997717.2 | 4 | 308 | |||
GenBank mRNA | BC017744.2 | - | 2 | 144 | ||
BC028282.1 | - | 3 | 179 | |||
BC042036.1 | - | 4 | 304 | |||
BU600037.1 | - | 313 | ||||
DQ225187.1 | - | 4 | 270 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
