siRNA Details
Gene Symbol: BOLA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035505.1 | NP_001030582.1 | 3 | 352 | ||
NM_212552.2 | NP_997717.2 | 4 | 441 | |||
GenBank mRNA | BC017744.2 | - | 2 | 277 | ||
BC028282.1 | - | 3 | 312 | |||
BC042036.1 | - | 4 | 437 | |||
BU600037.1 | - | 446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
