siRNA Details
Gene Symbol: PSG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC020759.1 | - | 4 | 1032 | 4731739 |
Gene Symbol: PSG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021016.3 | NP_066296.2 | 4 | 1067 | ||
GenBank mRNA | BC005924.1 | - | 4 | 1032 | 4043628 | |
M23575.1 | - | 4 | 1166 | |||
M27907.1 | - | 1 | 151 | |||
M30628.1 | - | 6 | 1256 | |||
M34420.1 | - | 4 | 1016 | |||
M37399.1 | - | 4 | 1021 | |||
M93061.1 | - | 4 | 993 | |||
S59493.1 | - | 4 | 993 | |||
S59494.1 | - | 6 | 1262 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
