siRNA Details
Gene Symbol: PSG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021016.3 | NP_066296.2 | 2 | 395 | ||
XM_011527126.1 | XP_011525428.1 | 2 | 395 | |||
XM_011527127.1 | XP_011525429.1 | 2 | 395 | |||
GenBank mRNA | BC005924.1 | - | 2 | 360 | 4043628 | |
DA833589.1 | - | 2 | 395 | |||
M23575.1 | - | 2 | 494 | |||
M30628.1 | - | 3 | 244 | |||
M34420.1 | - | 2 | 344 | |||
M37399.1 | - | 2 | 349 | |||
M93061.1 | - | 2 | 321 | |||
S59493.1 | - | 2 | 321 | |||
S59494.1 | - | 3 | 250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
