siRNA Details
Gene Symbol: PSG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021016.3 | NP_066296.2 | 2 | 423 | ||
XM_011527126.1 | XP_011525428.1 | 2 | 423 | |||
XM_011527127.1 | XP_011525429.1 | 2 | 423 | |||
GenBank mRNA | BC005924.1 | - | 2 | 388 | 4043628 | |
DA833589.1 | - | 2 | 423 | |||
M23575.1 | - | 2 | 522 | |||
M30628.1 | - | 3 | 272 | |||
M34420.1 | - | 2 | 372 | |||
M37399.1 | - | 2 | 377 | |||
M93061.1 | - | 2 | 349 | |||
S59493.1 | - | 2 | 349 | |||
S59494.1 | - | 3 | 278 |
Gene Symbol: PSG10P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026824.1 | 2 | 390 | |||
GenBank mRNA | AK309876.1 | - | 2 | 390 |
Gene Symbol: PSG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002785.2 | NP_002776.3 | 2 | 398 | ||
GenBank mRNA | BF084984.1 | - | 1 | 210 | ||
BG619770.1 | - | 2 | 399 | |||
M69245.1 | - | 2 | 360 |
Gene Symbol: PSG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002783.2 | NP_002774.2 | 2 | 393 | ||
GenBank mRNA | BX952498.1 | - | 2 | 391 | ||
U18467.1 | - | 2 | 343 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
