siRNA Details
Gene Symbol: HIST2H2AA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003516.2 | NP_003507.1 | 1 | 78 | ||
GenBank mRNA | AK312163.1 | - | 1 | 77 | ||
BC001629.1 | - | 1 | 63 | |||
BC019308.1 | - | 1 | 63 | |||
CR536525.1 | - | 1 | 25 | |||
CR541872.1 | - | 1 | 25 | |||
L19779.1 | - | 1 | 78 |
Gene Symbol: HIST2H2AA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040874.1 | NP_001035807.1 | 1 | 78 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013549, NM_178212, BC010564, AK002725, BC064002, and others.
Gene Ontology Categories:
