siRNA Details
Gene Symbol: ATP8A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313741.1 | NP_001300670.1 | 21 | 1829 | ||
NM_016529.5 | NP_057613.4 | 21 | 2040 | |||
XM_005266419.1 | XP_005266476.1 | 21 | 1794 | |||
XM_011535103.1 | XP_011533405.1 | 21 | 2040 | |||
XM_011535104.2 | XP_011533406.1 | 21 | 1816 | |||
XM_011535106.1 | XP_011533408.1 | 21 | 2040 | |||
XM_011535107.2 | XP_011533409.1 | 21 | 2040 | |||
XM_011535109.2 | XP_011533411.1 | 20 | 2326 | |||
XM_011535112.1 | XP_011533414.1 | 20 | 1637 | |||
XM_011535113.2 | XP_011533415.1 | 21 | 2040 | |||
XM_017020625.1 | XP_016876114.1 | 21 | 2040 | |||
XM_017020626.1 | XP_016876115.1 | 21 | 2040 | |||
GenBank mRNA | AK094653.1 | - | 10 | 765 | ||
AK126031.1 | - | 15 | 1253 | |||
AK127263.1 | - | 21 | 1980 | |||
AK302980.1 | - | 20 | 1775 | |||
AK310254.1 | - | 6 | 589 | |||
AL137256.1 | - | 7 | 462 | |||
BC144229.1 | - | 20 | 1750 | |||
BX537836.1 | - | 14 | 1258 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
