siRNA Details
Gene Symbol: NPIPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006985.2 | NP_008916.2 | 5 | 510 | ||
XM_011522748.2 | XP_011521050.1 | 5 | 607 | |||
XM_011522749.2 | XP_011521051.1 | 6 | 719 | |||
XM_011522751.1 | XP_011521053.1 | 5 | 606 | |||
XM_017023891.1 | XP_016879380.1 | 6 | 719 | |||
GenBank mRNA | AF132984.1 | - | 5 | 510 | ||
AK160377.1 | - | 24 | 4697 | |||
KF175165.1 | - | 5 | 546 | |||
KF175167.1 | - | 5 | 546 | |||
KF175169.1 | - | 5 | 510 | |||
KF175208.1 | - | 5 | 657 | |||
KF175209.1 | - | 6 | 728 | |||
KF175225.1 | - | 5 | 510 |
Gene Symbol: LOC613037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023520.1 | XP_016879009.1 | 1 | 1978 |
Gene Symbol: NPIPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | KF175213.1 | - | 4 | 203 |
Gene Symbol: NPIPA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277325.1 | NP_001264254.1 | 5 | 510 | ||
XM_011522329.2 | XP_011520631.1 | 7 | 921 | |||
XM_011522336.2 | XP_011520638.1 | 7 | 914 | |||
GenBank mRNA | CB123100.1 | - | 2 | 188 | ||
EL734608.1 | - | 5 | 510 | |||
KF175168.1 | - | 5 | 507 | |||
KF175170.1 | - | 5 | 510 | |||
KF175171.1 | - | 5 | 504 | |||
KF175172.1 | - | 5 | 510 | |||
KF175173.1 | - | 3 | 285 |
Gene Symbol: NPIPA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282507.1 | NP_001269436.1 | 5 | 510 | ||
XM_005255735.4 | XP_005255792.1 | 5 | 632 | |||
XM_011522339.2 | XP_011520641.1 | 5 | 558 | |||
XM_017022833.1 | XP_016878322.1 | 6 | 583 | |||
XM_017022834.1 | XP_016878323.1 | 6 | 635 | |||
XM_017022835.1 | XP_016878324.1 | 6 | 696 | |||
XM_017022836.1 | XP_016878325.1 | 5 | 503 | |||
XM_017022837.1 | XP_016878326.1 | 6 | 583 | |||
XM_017022838.1 | XP_016878327.1 | 5 | 978 | |||
XM_017022840.1 | XP_016878329.1 | 5 | 523 | |||
GenBank mRNA | BI046418.1 | - | 2 | 161 |
Gene Symbol: NPIPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282511.1 | NP_001269440.1 | 5 | 510 | ||
XM_005255738.4 | XP_005255795.1 | 5 | 632 | |||
XM_006720830.3 | XP_006720893.1 | 5 | 1530 | |||
XM_011522342.2 | XP_011520644.1 | 5 | 559 | |||
XM_017022841.1 | XP_016878330.1 | 6 | 583 | |||
XM_017022842.1 | XP_016878331.1 | 6 | 635 | |||
XM_017022843.1 | XP_016878332.1 | 6 | 696 | |||
XM_017022844.1 | XP_016878333.1 | 5 | 503 | |||
XM_017022845.1 | XP_016878334.1 | 6 | 583 | |||
XM_017022847.1 | XP_016878336.1 | 5 | 523 | |||
GenBank mRNA | BY996642.1 | - | 1 | 91 |
Gene Symbol: NPIPB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011523501.2 | XP_011521803.1 | 1 | 32 |
Gene Symbol: NPIPB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128772.1 | - | 1 | 697 |
Gene Symbol: NPIPB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005255743.3 | XP_005255800.1 | 1 | 1993 |
Gene Symbol: NPIPB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022827.1 | XP_016878316.1 | 1 | 460 |
Gene Symbol: PDXDC2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX647358.1 | - | 1 | 2833 |
Gene Symbol: PKD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK298381.1 | - | 3 | 268 | ||
BC039707.1 | - | 6 | 829 | 4479933 | ||
BC046145.1 | - | 7 | 921 | |||
KF175174.1 | - | 5 | 546 | |||
KF175210.1 | - | 6 | 792 | |||
KF175212.1 | - | 8 | 679 | |||
KF175214.1 | - | 6 | 728 | |||
KF175219.1 | - | 5 | 696 |
Gene Symbol: PKD1P6-NPIPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_123722.1 | 14 | 2464 | |||
GenBank mRNA | AK131085.1 | - | 18 | 4873 | ||
AK302708.1 | - | 14 | 2464 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
