siRNA Details
Gene Symbol: POTEB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207355.3 | NP_997238.2 | 1 | 624 | ||
NR_110752.1 | 1 | 624 | ||||
XM_006720358.2 | XP_006720421.1 | 1 | 671 | |||
XM_011543795.2 | XP_011542097.1 | 1 | 671 | |||
XM_011543796.2 | XP_011542098.1 | 1 | 671 | |||
XM_011543797.2 | XP_011542099.1 | 1 | 671 | |||
GenBank mRNA | AY465173.1 | - | 1 | 301 | ||
BC066981.1 | - | 1 | 624 | 4824630 | ||
BC101006.1 | - | 1 | 286 | 40006485 | ||
BC127123.1 | - | 1 | 286 | 40006490 |
Gene Symbol: LOC102723502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724900.3 | XP_006724963.1 | 1 | 400 | ||
XM_006724901.3 | XP_006724964.1 | 1 | 400 | |||
XM_011546199.2 | XP_011544501.1 | 1 | 400 | |||
XM_011546200.2 | XP_011544502.1 | 1 | 400 |
Gene Symbol: POTEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137671.1 | NP_001131143.1 | 1 | 707 | ||
NR_134878.1 | 1 | 707 | ||||
GenBank mRNA | AK307658.1 | - | 1 | 707 | ||
BX649118.1 | - | 1 | 693 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
