siRNA Details
Gene Symbol: POTEB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207355.3 | NP_997238.2 | 4 | 1269 | ||
NR_110752.1 | 4 | 1269 | ||||
XM_006720358.2 | XP_006720421.1 | 4 | 1316 | |||
XM_011543795.2 | XP_011542097.1 | 4 | 1316 | |||
XM_011543796.2 | XP_011542098.1 | 4 | 1316 | |||
XM_011543797.2 | XP_011542099.1 | 4 | 1316 | |||
GenBank mRNA | AY465170.1 | - | 5 | 839 | ||
AY465173.1 | - | 4 | 946 | |||
BC066981.1 | - | 4 | 1269 | 4824630 | ||
BC101005.1 | - | 4 | 820 | 40006483 | ||
BC101006.1 | - | 4 | 931 | 40006485 | ||
BC127123.1 | - | 4 | 931 | 40006490 |
Gene Symbol: LOC102723502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006724900.3 | XP_006724963.1 | 4 | 1045 | ||
XM_006724901.3 | XP_006724964.1 | 4 | 1045 | |||
XM_011546199.2 | XP_011544501.1 | 4 | 1045 | |||
XM_011546200.2 | XP_011544502.1 | 4 | 1045 |
Gene Symbol: POTEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002920.1 | NP_001002920.1 | 4 | 719 | ||
NM_001005365.2 | NP_001005365.2 | 4 | 719 | |||
XM_011544516.1 | XP_011542818.1 | 4 | 791 | |||
GenBank mRNA | AY462869.1 | - | 4 | 719 | ||
AY462870.1 | - | 4 | 719 |
Gene Symbol: POTEB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277304.1 | NP_001264233.1 | 4 | 820 | ||
NR_102391.1 | 4 | 820 | ||||
XM_006720355.3 | XP_006720418.1 | 4 | 934 | |||
XM_011543794.2 | XP_011542096.1 | 4 | 934 | |||
XM_017021858.1 | XP_016877347.1 | 4 | 934 |
Gene Symbol: POTEB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277303.1 | NP_001264232.1 | 4 | 820 | ||
NR_102390.1 | 4 | 820 | ||||
XM_006720354.3 | XP_006720417.1 | 4 | 934 | |||
XM_011543793.2 | XP_011542095.1 | 4 | 934 | |||
XM_017021853.1 | XP_016877342.1 | 4 | 934 | |||
GenBank mRNA | BC101007.1 | - | 4 | 820 | 40006489 |
Gene Symbol: POTEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137671.1 | NP_001131143.1 | 4 | 1352 | ||
NR_134878.1 | 4 | 1352 | ||||
GenBank mRNA | AK307658.1 | - | 4 | 1352 | ||
BC140940.1 | - | 4 | 1245 | 9056752 | ||
BX649118.1 | - | 4 | 1338 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
