siRNA Details
Gene Symbol: UPF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023011.3 | NP_075387.1 | 3 | 397 | ||
NM_080687.2 | NP_542418.1 | 3 | 397 | |||
XM_006719991.3 | XP_006720054.1 | 2 | 229 | |||
XM_011534844.1 | XP_011533146.1 | 3 | 375 | |||
XM_011534845.2 | XP_011533147.1 | 3 | 882 | |||
XM_011534846.1 | XP_011533148.1 | 3 | 344 | |||
XM_011534847.2 | XP_011533149.1 | 1 | 472 | |||
XM_011534848.2 | XP_011533150.1 | 3 | 882 | |||
XM_017020709.1 | XP_016876198.1 | 2 | 1066 | |||
XM_017020710.1 | XP_016876199.1 | 2 | 229 | |||
XM_017020711.1 | XP_016876200.1 | 2 | 229 | |||
XM_017020712.1 | XP_016876201.1 | 3 | 390 | |||
GenBank mRNA | AF318575.1 | - | 3 | 290 | ||
AK294545.1 | - | 3 | 344 | |||
AK302003.1 | - | 3 | 333 | |||
AY013250.1 | - | 3 | 397 | |||
BC023569.1 | - | 3 | 361 | 4127465 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
