siRNA Details
Gene Symbol: UPF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080687.2 | NP_542418.1 | 3, 4 | 478 | ||
XM_017020712.1 | XP_016876201.1 | 3, 4 | 471 | |||
GenBank mRNA | BC023569.1 | - | 3, 4 | 442 | 4127465 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
