siRNA Details
Gene Symbol: DEXI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014015.3 | NP_054734.2 | 1 | 513 | ||
XM_017023171.1 | XP_016878660.1 | 1 | 1120 | |||
GenBank mRNA | AF108145.2 | - | 1 | 129 | ||
AF129929.1 | - | 1 | 25 | |||
AK314111.1 | - | 1 | 482 | |||
BC001083.2 | - | 1 | 249 | 2958243 | ||
BC037569.1 | - | 1 | 513 | 5227606 | ||
BC052989.1 | - | 1 | 246 | 6194689 | ||
BC071683.1 | - | 1 | 412 | 5190179 |
Gene Symbol: LOC100289656 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036475.2 | 2 | 819 | |||
GenBank mRNA | AK302529.1 | - | 2 | 819 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
