siRNA Details
Gene Symbol: OR4C46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004703.1 | NP_001004703.1 | 1 | 467 |
Gene Symbol: OR4C13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001955.2 | NP_001001955.2 | 1 | 499 | ||
GenBank mRNA | BC137005.1 | - | 1 | 499 | 9020998 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
