siRNA Details
Gene Symbol: SPDYE16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006716211.3 | XP_006716274.1 | 2 | 587 | ||
XM_017012887.1 | XP_016868376.1 | 2 | 587 |
Gene Symbol: SPDYE18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250742.4 | XP_005250799.1 | 3 | 458 | ||
XM_005250743.4 | XP_005250800.1 | 2 | 313 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
