siRNA Details
Gene Symbol: LOC101060524 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111000.1 | 1 | 1044 |
Gene Symbol: DRD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000798.4 | NP_000789.1 | 1 | 935 | ||
GenBank mRNA | AK313897.1 | - | 1 | 762 | ||
AY136750.1 | - | 1 | 539 | |||
BC009748.2 | - | 1 | 590 | 3928370 |
Gene Symbol: DRD5P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_111001.1 | 1 | 1044 | |||
GenBank mRNA | AK225830.1 | - | 1 | 1044 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
