siRNA Details
Gene Symbol: ATP5J2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003713.2 | NP_001003713.1 | 2 | 101 | ||
NM_001003714.2 | NP_001003714.1 | 2 | 119 | |||
NM_001039178.2 | NP_001034267.1 | 2 | 101 | |||
NM_004889.3 | NP_004880.1 | 2 | 119 | |||
GenBank mRNA | AF047436.1 | - | 2 | 86 | ||
AF088918.1 | - | 2 | 72 | |||
AK123419.1 | - | 1 | 1323 | |||
AK223348.1 | - | 2 | 82 | |||
AY046911.1 | - | 2 | 61 | |||
BU587946.1 | - | 2 | 89 | |||
CB110074.1 | - | 2 | 86 | |||
CB155273.1 | - | 2 | 86 | |||
CD173604.1 | - | 2 | 123 | |||
CD176299.1 | - | 2 | 81 | |||
CN274665.1 | - | 2 | 53 | |||
CR456891.1 | - | 2 | 59 | |||
CR542155.1 | - | 2 | 59 | |||
DA064847.1 | - | 2 | 82 | |||
DB114195.1 | - | 2 | 86 | |||
EL952520.1 | - | 2 | 73 |
Gene Symbol: ATP5J2-PTCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198879.1 | NP_001185808.1 | 2 | 101 | ||
GenBank mRNA | AK295912.1 | - | 2 | 64 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
