siRNA Details
Gene Symbol: ATP5J2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003713.2 | NP_001003713.1 | 1, 2 | 78 | ||
NM_001039178.2 | NP_001034267.1 | 1, 2 | 78 | |||
GenBank mRNA | AY046911.1 | - | 1, 2 | 38 | ||
BU587946.1 | - | 1, 2 | 66 | |||
CB155273.1 | - | 1, 2 | 63 | |||
CD173604.1 | - | 1, 2 | 100 | |||
CD176299.1 | - | 1, 2 | 58 | |||
CN274665.1 | - | 1, 2 | 30 | |||
DB114195.1 | - | 1, 2 | 63 |
Gene Symbol: ATP5J2-PTCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198879.1 | NP_001185808.1 | 1, 2 | 78 | ||
GenBank mRNA | AK295912.1 | - | 1, 2 | 41 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
