siRNA Details
Gene Symbol: ZNF695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204221.1 | NP_001191150.1 | 3, 4 | 452 | ||
NM_020394.4 | NP_065127.4 | 3, 4 | 452 | |||
NR_037892.1 | 3, 4 | 452 | ||||
GenBank mRNA | CN394881.1 | - | 3, 4 | 384 |
Gene Symbol: ZNF670-ZNF695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037894.1 | 3, 4 | 448 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
