siRNA Details
Gene Symbol: ZNF695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204221.1 | NP_001191150.1 | 4 | 502 | ||
NM_020394.4 | NP_065127.4 | 4 | 502 | |||
NR_037892.1 | 4 | 502 | ||||
GenBank mRNA | BC023527.2 | - | 4 | 410 | ||
BC041082.1 | - | 4 | 439 | |||
BE743952.1 | - | 3 | 241 | |||
CN394881.1 | - | 4 | 434 | |||
DA809929.1 | - | 4 | 502 |
Gene Symbol: ZNF670-ZNF695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037894.1 | 4 | 498 | |||
GenBank mRNA | AK126366.1 | - | 6 | 657 | ||
AL524527.3 | - | 4 | 421 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
