siRNA Details
Gene Symbol: ZNF695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204221.1 | NP_001191150.1 | 2 | 227 | ||
NM_020394.4 | NP_065127.4 | 2 | 227 | |||
NR_037892.1 | 2 | 227 | ||||
GenBank mRNA | BC023527.2 | - | 2 | 135 | ||
BC041082.1 | - | 2 | 164 | |||
BC055096.1 | - | 2 | 191 | |||
CN394881.1 | - | 2 | 159 | |||
DA809929.1 | - | 2 | 227 |
Gene Symbol: ZNF670-ZNF695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037894.1 | 2 | 259 | |||
GenBank mRNA | AK126366.1 | - | 2 | 257 | ||
AL524527.3 | - | 2 | 182 |
Gene Symbol: ZNF678 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178549.3 | NP_848644.2 | 3 | 338 | ||
NR_033184.1 | 2 | 245 | ||||
NR_102302.1 | 3 | 338 | ||||
GenBank mRNA | AK307248.1 | - | 3 | 314 | ||
BC042500.1 | - | 2 | 221 | 4823179 | ||
CD655825.1 | - | 3 | 273 | |||
DA213851.1 | - | 2 | 246 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
