siRNA Details
Gene Symbol: RPL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000977.3 | NP_000968.2 | 3 | 272 | ||
NM_001243130.1 | NP_001230059.1 | 3 | 272 | |||
NM_001243131.1 | NP_001230060.1 | 3 | 272 | |||
NM_033251.2 | NP_150254.1 | 2 | 445 | |||
GenBank mRNA | AB062392.1 | - | 2 | 221 | ||
AK025405.1 | - | 1 | 266 | |||
AK026501.1 | - | 1 | 657 | |||
AK127579.1 | - | 3 | 249 | |||
AK130963.1 | - | 3 | 168 | |||
AK290893.1 | - | 3 | 247 | |||
AK297198.1 | - | 3 | 248 | |||
BC000851.1 | - | 1 | 225 | |||
BC004954.1 | - | 1 | 235 | 3623320 | ||
BC006179.1 | - | 3 | 235 | |||
BC007345.2 | - | 3 | 237 | 3688320 | ||
BC007563.1 | - | 2 | 433 | 2989021 | ||
BC007805.2 | - | 3 | 233 | 4298327 | ||
BC010994.2 | - | 2 | 228 | 3842093 | ||
BC013078.1 | - | 3 | 233 | 3459415 | ||
BC014167.2 | - | 1 | 233 | 4550361 | ||
BC020804.1 | - | 2 | 445 | 4276103 | ||
BC027463.1 | - | 3 | 233 | 5111302 | ||
BC063378.1 | - | 1 | 236 | 3542514 | ||
BC066320.1 | - | 3 | 247 | 5300601 | ||
BC093063.1 | - | 1 | 249 | 30401947 | ||
BC106058.1 | - | 3 | 272 | 4992467 | ||
X64707.1 | - | 3 | 247 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
